Product Details

SNP ID
rs10795929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12081772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGGGGCATGTTGAAGTACGGTC[A/C]GGGAGGAGCATTCTTGGGATTCTAG
Phenotype
MIM: 614984
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DHTKD1 PubMed Links

Gene Details

Gene
DHTKD1
Gene Name
dehydrogenase E1 and transketolase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018706.6 Intron NP_061176.3

View Full Product Details