Product Details

SNP ID
rs61752753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10972348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGATGTACATTTATACTAAAAAA[A/C]CAAAAACAAAGTTTGTACAAGATGA
Phenotype
MIM: 604275
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CTNND2 PubMed Links

Gene Details

Gene
CTNND2
Gene Name
catenin delta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288715.1 4043 UTR 3 NP_001275644.1
NM_001288716.1 4043 UTR 3 NP_001275645.1
NM_001288717.1 4043 UTR 3 NP_001275646.1
NM_001332.3 4043 UTR 3 NP_001323.1
XM_005248251.3 4043 Intron XP_005248308.1
XM_005248252.2 4043 Intron XP_005248309.1
XM_005248253.1 4043 Intron XP_005248310.1
XM_011513967.2 4043 Intron XP_011512269.1
XM_017009072.1 4043 Intron XP_016864561.1
XM_017009073.1 4043 Intron XP_016864562.1
XM_017009074.1 4043 Intron XP_016864563.1
XM_017009075.1 4043 Intron XP_016864564.1
XM_017009076.1 4043 Intron XP_016864565.1

View Full Product Details