Product Details

SNP ID
rs6882
Assay Type
Functionally tested
NCBI dbSNP Submissions
71
Location
Chr.1:42746440 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AGACACTTAAGTACTGTATCGCTGT[C/T]ATGCAGCGGCCTGTGGAGGCCCCTG
Phenotype
MIM: 610036 MIM: 610339
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN19 PubMed Links

Gene Details

Gene
CLDN19
Gene Name
claudin 19
There are no transcripts associated with this gene.

Gene
P3H1
Gene Name
prolyl 3-hydroxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146289.1 2550 UTR 3 NP_001139761.1
NM_001243246.1 2550 UTR 3 NP_001230175.1
NM_022356.3 2550 UTR 3 NP_071751.3
XM_005271110.2 2550 UTR 3 XP_005271167.1
XM_011541948.1 2550 UTR 3 XP_011540250.1
XM_017002051.1 2550 UTR 3 XP_016857540.1
XM_017002052.1 2550 UTR 3 XP_016857541.1

View Full Product Details