Product Details

SNP ID
rs17017450
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:211383820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTAAGCTCTAAACTTAATAATAT[G/T]TTGCATATCCAAAATGTGCTCATGC
Phenotype
MIM: 602356
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00467 PubMed Links

Gene Details

Gene
LINC00467
Gene Name
long intergenic non-protein coding RNA 467
There are no transcripts associated with this gene.

Gene
TRAF5
Gene Name
TNF receptor associated factor 5
There are no transcripts associated with this gene.

View Full Product Details