Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_030625.2 | 367 | UTR 5 | NP_085128.2 | ||
XM_011540204.2 | 367 | UTR 5 | XP_011538506.1 | ||
XM_011540205.2 | 367 | Intron | XP_011538507.1 | ||
XM_011540206.2 | 367 | UTR 5 | XP_011538508.1 | ||
XM_011540207.2 | 367 | Intron | XP_011538509.1 | ||
XM_017016686.1 | 367 | Intron | XP_016872175.1 | ||
XM_017016687.1 | 367 | Intron | XP_016872176.1 | ||
XM_017016688.1 | 367 | Intron | XP_016872177.1 | ||
XM_017016689.1 | 367 | Intron | XP_016872178.1 |