Product Details

SNP ID
rs10828993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:26442199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAACCAGGGTCACGGGAGAAGG[A/T]TGAATAGAAGGAAACCAAGCAAAGA
Phenotype
MIM: 609036
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
APBB1IP PubMed Links

Gene Details

Gene
APBB1IP
Gene Name
amyloid beta precursor protein binding family B member 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019043.3 Intron NP_061916.3
XM_006717451.2 Intron XP_006717514.1
XM_011519514.2 Intron XP_011517816.1

View Full Product Details