Product Details

SNP ID
rs4499592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129432204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGGTGTACTTTTGACAGGAGGA[A/C]AACCACGTCAGGCTTAGAGATAGCA
Phenotype
MIM: 606045 MIM: 603574
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB12 PubMed Links

Gene Details

Gene
EFCAB12
Gene Name
EF-hand calcium binding domain 12
There are no transcripts associated with this gene.

Gene
IFT122
Gene Name
intraflagellar transport 122
There are no transcripts associated with this gene.

Gene
MBD4
Gene Name
methyl-CpG binding domain 4, DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276270.1 2310 Intron NP_001263199.1
NM_001276271.1 2310 UTR 3 NP_001263200.1
NM_001276272.1 2310 Intron NP_001263201.1
NM_001276273.1 2310 Intron NP_001263202.1
NM_003925.2 2310 Intron NP_003916.1

View Full Product Details