Product Details

SNP ID
rs2555691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54586068 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAATTTTAGAAATCATAATACATG[A/G]AAATTTTATTTCTTTATTTCTAAAT
Phenotype
MIM: 604810 MIM: 604812
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LILRA1 PubMed Links

Gene Details

Gene
LILRA1
Gene Name
leukocyte immunoglobulin like receptor A1
There are no transcripts associated with this gene.

Gene
LILRA2
Gene Name
leukocyte immunoglobulin like receptor A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130917.2 Intron NP_001124389.2
NM_001290270.1 Intron NP_001277199.1
NM_001290271.1 Intron NP_001277200.1
NM_006866.3 Intron NP_006857.2
XM_006722986.1 Intron XP_006723049.1
XM_011526385.1 Intron XP_011524687.1
XM_011526386.1 Intron XP_011524688.1
XM_011526387.1 Intron XP_011524689.1
XM_011526388.1 Intron XP_011524690.1
XM_011526389.1 Intron XP_011524691.1
XM_011526390.1 Intron XP_011524692.1
XM_011526391.1 Intron XP_011524693.1
XM_017026224.1 Intron XP_016881713.1

View Full Product Details