Product Details

SNP ID
rs34895155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31078670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCTCTGTTTATCTGAGTAATAG[C/T]CACTTCCTCTATTAAAGTCTTTTTT
Phenotype
MIM: 601150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX11 PubMed Links

Gene Details

Gene
DDX11
Gene Name
DEAD/H-box helicase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257144.1 Intron NP_001244073.1
NM_001257145.1 Intron NP_001244074.1
NM_004399.2 Intron NP_004390.3
NM_030653.3 Intron NP_085911.2
NM_152438.1 Intron NP_689651.1
XM_005253331.3 Intron XP_005253388.1
XM_005253333.3 Intron XP_005253390.1
XM_006719050.3 Intron XP_006719113.1
XM_011520592.2 Intron XP_011518894.1
XM_011520595.2 Intron XP_011518897.1
XM_011520597.2 Intron XP_011518899.1
XM_017018918.1 Intron XP_016874407.1
XM_017018919.1 Intron XP_016874408.1
XM_017018920.1 Intron XP_016874409.1
XM_017018921.1 Intron XP_016874410.1
XM_017018922.1 Intron XP_016874411.1
XM_017018923.1 Intron XP_016874412.1
XM_017018924.1 Intron XP_016874413.1
XM_017018925.1 Intron XP_016874414.1
XM_017018926.1 Intron XP_016874415.1
XM_017018927.1 Intron XP_016874416.1
XM_017018928.1 Intron XP_016874417.1
XM_017018929.1 Intron XP_016874418.1
XM_017018930.1 Intron XP_016874419.1
XM_017018931.1 Intron XP_016874420.1
XM_017018932.1 Intron XP_016874421.1
XM_017018933.1 Intron XP_016874422.1
XM_017018934.1 Intron XP_016874423.1
XM_017018935.1 Intron XP_016874424.1
XM_017018936.1 Intron XP_016874425.1
XM_017018937.1 Intron XP_016874426.1
XM_017018938.1 Intron XP_016874427.1
XM_017018939.1 Intron XP_016874428.1
Gene
DDX11-AS1
Gene Name
DDX11 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details