Product Details

SNP ID
hCV26282821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:39148941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTATTTTTCCAGGTGTGACCCTA[T/C]GATTCTTCTCAGCCAAAATCTTTGT
Phenotype
MIM: 610517
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CNTNAP3 PubMed Links

Gene Details

Gene
CNTNAP3
Gene Name
contactin associated protein-like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033655.3 Intron NP_387504.2

View Full Product Details