Product Details
- SNP ID
-
rs11643752
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31440378 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCATTTTAACTACTCCATAATCATA[G/A]TTTAGACACAGAGCAAGAGGAAGCC
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF843
PubMed Links
Gene Details
- Gene
- ZNF843
- Gene Name
- zinc finger protein 843
View Full Product Details