Product Details

SNP ID
rs1122186
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:153932990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGATTTCCTCATCATACACCAGC[G/T]AATCACAGGCACGGCACAGGGAGCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DENND4B PubMed Links

Gene Details

Gene
DENND4B
Gene Name
DENN domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014856.2 3895 Nonsense Mutation TAG,TCG *1165S NP_055671.2

View Full Product Details