Product Details

SNP ID
rs799906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43126099 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCAGGAGTCTGGGGCAAGTAGTC[C/T]TGTAAGGTCAGTGGCCTGCGGGGAC
Phenotype
MIM: 113705
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRCA1 PubMed Links

Gene Details

Gene
BRCA1
Gene Name
BRCA1, DNA repair associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007294.3 Intron NP_009225.1
NM_007297.3 Intron NP_009228.2
NM_007298.3 Intron NP_009229.2
NM_007299.3 Intron NP_009230.2
NM_007300.3 Intron NP_009231.2
Gene
NBR2
Gene Name
neighbor of BRCA1 gene 2 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details