Product Details
- SNP ID
-
rs1951630
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:102610470 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTGCCATATTGGACAGCTCAGAAT[A/T]GAGTATTTTTATCCTCCCAGCAAGT
- Phenotype
-
MIM: 607675
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RCOR1
PubMed Links
Gene Details
- Gene
- RCOR1
- Gene Name
- REST corepressor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015156.3 |
|
Intron |
|
|
NP_055971.2 |
View Full Product Details