Product Details

SNP ID
hCV26381064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:77693297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTTGAAGATCCAAATATGCGGCT[C/T]CCAACCTGCGCTGGCAATCTGGGAT
Phenotype
MIM: 610058
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBCA PubMed Links

Gene Details

Gene
TBCA
Gene Name
tubulin folding cofactor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297738.1 319 Missense Mutation GAA,GGA E72G NP_001284667.1
NM_001297740.1 319 Intron NP_001284669.1
NM_004607.2 319 Missense Mutation GAA,GGA E72G NP_004598.1

View Full Product Details