Product Details

SNP ID
rs1047726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:169729589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGCCACCTTACTCATGCAGCAA[A/G]TTGGTCTATGTTCTCTTGACTTCTG
Phenotype
MIM: 600580 MIM: 616614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCN3 PubMed Links

Gene Details

Gene
CLCN3
Gene Name
chloride voltage-gated channel 3
There are no transcripts associated with this gene.

Gene
HPF1
Gene Name
histone PARylation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017867.2 Intron NP_060337.2
XM_017008332.1 Intron XP_016863821.1

View Full Product Details