Product Details
- SNP ID
-
rs2169836
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:70296684 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGATGACCAGAAATGGGGATCCAC[A/T]AGGAACATTTCAAAGTTCTTATCCA
- Phenotype
-
MIM: 616275
MIM: 603542
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM136A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145815815] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM136A
- Gene Name
- family with sequence similarity 136 member A
- Gene
- SNRPG
- Gene Name
- small nuclear ribonucleoprotein polypeptide G
There are no transcripts associated with this gene.
View Full Product Details