Product Details

SNP ID
rs2604978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:97925613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCACACTCCTCACCACAGGCCAC[A/C]TTTTTTGTTTTTATTTTTTTTTTTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01550 PubMed Links

Gene Details

Gene
LINC01550
Gene Name
long intergenic non-protein coding RNA 1550
There are no transcripts associated with this gene.

View Full Product Details