Product Details

SNP ID
rs11761180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16464081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGGGACACTGAAGTGGTTCCAC[A/G]AAAAGTACTGAAAGATACGTCGAAG
Phenotype
MIM: 609675
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SOSTDC1 PubMed Links

Gene Details

Gene
SOSTDC1
Gene Name
sclerostin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015464.2 Intron NP_056279.1
XM_017011943.1 Intron XP_016867432.1

View Full Product Details