Product Details

SNP ID
rs15504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16791913 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTCCAGGGGCCAAAGAAATCAG[G/T]AGGGGCTGGGCAGTAGAGGAATTCC
Phenotype
MIM: 606358 MIM: 613139
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AGR2 PubMed Links

Gene Details

Gene
AGR2
Gene Name
anterior gradient 2, protein disulphide isomerase family member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006408.3 1679 Intron NP_006399.1
XM_005249581.4 1679 UTR 3 XP_005249638.1
Gene
TSPAN13
Gene Name
tetraspanin 13
There are no transcripts associated with this gene.

View Full Product Details