Product Details

SNP ID
rs11095518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:9731801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTGGGCAATATAAGGAATTGGG[A/G]GGGGGGGGAAGGAATTGTGGAAGGC
Phenotype
MIM: 300808
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR143 PubMed Links

Gene Details

Gene
GPR143
Gene Name
G protein-coupled receptor 143
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000273.2 Intron NP_000264.2
XM_005274541.3 Intron XP_005274598.1

View Full Product Details