Product Details

SNP ID
rs4622271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66745233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGAGGTGCCTGCCGGAGCCCAG[A/C]GCCTTCCATGCACCGGGTGCTGGGA
Phenotype
MIM: 616109
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C11orf80 PubMed Links

Gene Details

Gene
C11orf80
Gene Name
chromosome 11 open reading frame 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302084.1 Intron NP_001289013.1
NM_024650.3 Intron NP_078926.3

View Full Product Details