Product Details
- SNP ID
-
rs2514034
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:67644843 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACGACGCCCACGGGTCTATAGGCTT[A/C]CATCTCAAAGGCAGGAAAGGCCGTA
- Phenotype
-
MIM: 614413
MIM: 604648
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ACY3
PubMed Links
Gene Details
- Gene
- ACY3
- Gene Name
- aminoacylase 3
- Gene
- TBX10
- Gene Name
- T-box 10
There are no transcripts associated with this gene.
View Full Product Details