Product Details

SNP ID
rs1213585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85683991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAGGTATTCATAATAATGTTTA[C/T]ATATCAATACCTAATTACAGGAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC89 PubMed Links

Gene Details

Gene
CCDC89
Gene Name
coiled-coil domain containing 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152723.2 2287 UTR 3 NP_689936.1

View Full Product Details