Product Details
- SNP ID
-
rs35315047
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:90406831 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGACATTATAAAGGAAGATCAAAT[C/T]TGGGTAACCAAAAAAAAAAAAAAAC
- Phenotype
-
MIM: 602907
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CETN3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76863451] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CETN3
- Gene Name
- centrin 3
- Gene
- LOC731157
- Gene Name
- uncharacterized LOC731157
There are no transcripts associated with this gene.
View Full Product Details