Product Details
- SNP ID
-
rs7552561
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
31
- Location
-
Chr.1:229445239 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACATGCAAGAGACGCCAAGGAAAA[C/T]ACATTTAAAAAAAGATGTTATACTC
- Phenotype
-
MIM: 607613
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUP133
PubMed Links
Gene Details
- Gene
- NUP133
- Gene Name
- nucleoporin 133
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018230.2 |
|
Intron |
|
|
NP_060700.2 |
View Full Product Details