Product Details

SNP ID
rs5981583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:74084184 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGTGAATAATGATAAGTAAATT[A/T]TCAAATCAAAGTGTATGATCTTGCT
Phenotype
MIM: 300936
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FTX PubMed Links

Gene Details

Gene
FTX
Gene Name
FTX transcript, XIST regulator (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details