Product Details

SNP ID
hCV26622988
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8224408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACGTGGCTACACAGGACTCTAAG[C/T]AGAAAGGGAGGTTGCCCCCAAGAGT
Phenotype
MIM: 613041
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM86FP PubMed Links

Gene Details

Gene
FAM86FP
Gene Name
family with sequence similarity 86, member A pseudogene
There are no transcripts associated with this gene.

Gene
FAM90A1
Gene Name
family with sequence similarity 90 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319982.1 Intron NP_001306911.1
NM_018088.3 Intron NP_060558.3

View Full Product Details