Product Details

SNP ID
rs1011636963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75526417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGTGATAGGGGAAGCATGCAC[G/T]TGTAACTTTGTAACTCTAAGATAAT
Phenotype
MIM: 601767
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HIP1 PubMed Links

Gene Details

Gene
HIP1
Gene Name
huntingtin interacting protein 1
There are no transcripts associated with this gene.

Gene
PMS2P3
Gene Name
PMS1 homolog 2, mismatch repair system component pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details