Product Details

SNP ID
rs4244595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18774016 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACATACAGATGTAAGACTAGATC[A/G]GCTACTATTGATTACACTCTAATGG
Phenotype
MIM: 611679
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXW10 PubMed Links

Gene Details

Gene
FBXW10
Gene Name
F-box and WD repeat domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267585.1 Intron NP_001254514.1
NM_001267586.1 Intron NP_001254515.1
XM_011523625.1 Intron XP_011521927.1
XM_011523626.2 Intron XP_011521928.1
XM_017024020.1 Intron XP_016879509.1
XM_017024021.1 Intron XP_016879510.1
Gene
TVP23B
Gene Name
trans-golgi network vesicle protein 23 homolog B (S. cerevisiae)
There are no transcripts associated with this gene.

View Full Product Details