Product Details

SNP ID
rs2904802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:4967172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAACATAGAAGAAGAATCCTAAA[C/T]CTAACTCCTAATTCCTTTCTATGGG
Phenotype
MIM: 600449
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKR1C1 PubMed Links

Gene Details

Gene
AKR1C1
Gene Name
aldo-keto reductase family 1 member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001353.5 Intron NP_001344.2
XM_017015791.1 Intron XP_016871280.1

View Full Product Details