Product Details

SNP ID
hCV26668794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:94421006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATATAGTTAGAAAAACCAGTGGA[T/C]TAGCTGATATCTCAATACTTTATGC
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MFSD14B PubMed Links

Gene Details

Gene
MFSD14B
Gene Name
major facilitator superfamily domain containing 14B
There are no transcripts associated with this gene.

View Full Product Details