Product Details
- SNP ID
-
rs1802666
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:150796356 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAATAGATTTATGAATTGGAGTA[G/T]AAAACATAGACATTTCTACCTTGAG
- Phenotype
-
MIM: 601105
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CTSK
PubMed Links
Gene Details
- Gene
- CTSK
- Gene Name
- cathepsin K
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000396.3 |
1662 |
UTR 3 |
|
|
NP_000387.1 |
View Full Product Details