Product Details

SNP ID
rs3936009
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:1654188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAATTACTAATACCTGATGACG[T/G]ATACGAGCTAGATATGAGAAGAAAA
Phenotype
MIM: 176873
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
CDK11B PubMed Links

Gene Details

Gene
CDK11B
Gene Name
cyclin dependent kinase 11B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291345.1 Intron NP_001278274.1
NM_001787.2 Intron NP_001778.2
NM_033486.2 Intron NP_277021.2
NM_033487.2 Intron NP_277022.1
NM_033489.2 Intron NP_277024.2
NM_033490.2 Intron NP_277025.1
XM_006711065.3 Intron XP_006711128.1
XM_011542490.2 Intron XP_011540792.1
XM_011542491.2 Intron XP_011540793.1
XM_011542492.2 Intron XP_011540794.1
XM_011542493.2 Intron XP_011540795.1
XM_011542494.2 Intron XP_011540796.1
XM_011542495.2 Intron XP_011540797.1
XM_011542496.2 Intron XP_011540798.1
XM_017002925.1 Intron XP_016858414.1
XM_017002926.1 Intron XP_016858415.1
XM_017002927.1 Intron XP_016858416.1
XM_017002928.1 Intron XP_016858417.1
Gene
SLC35E2B
Gene Name
solute carrier family 35 member E2B
There are no transcripts associated with this gene.

View Full Product Details