Product Details

SNP ID
rs35941329
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:115286460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGCTCCTGTGAGTCCTGTTGAA[A/G]GGGGCAGCACCACCGACCTCGAAGT
Phenotype
MIM: 162030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NGF PubMed Links

Gene Details

Gene
NGF
Gene Name
nerve growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002506.2 505 Silent Mutation CCC,CCT P112P NP_002497.2
XM_006710663.3 505 Silent Mutation CCC,CCT P112P XP_006710726.1
XM_011541518.2 505 Silent Mutation CCC,CCT P167P XP_011539820.1

View Full Product Details