Product Details
- SNP ID
-
rs3926888
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:36763306 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAAGGGCTGCAAACGCCTGAAGGC[A/G]AGGGCCAAATTTATGTGAGCCTTTG
- Phenotype
-
MIM: 615870
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IFT27
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74494195] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFT27
- Gene Name
- intraflagellar transport 27
- Gene
- LOC105373021
- Gene Name
- uncharacterized LOC105373021
There are no transcripts associated with this gene.
View Full Product Details