Product Details

SNP ID
rs11666491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:512568 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGGCTCTTAACATTCCCGCTGAA[A/C]GTGGGCCACGCCCCCTCGCACGTTT
Phenotype
MIM: 102670
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MADCAM1 PubMed Links

Gene Details

Gene
MADCAM1
Gene Name
mucosal vascular addressin cell adhesion molecule 1
There are no transcripts associated with this gene.

Gene
TPGS1
Gene Name
tubulin polyglutamylase complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033513.2 Intron NP_277048.2

View Full Product Details