Product Details
- SNP ID
-
rs12984789
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:509365 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGAGCACTGCACAAGCCACTCAC[C/T]CCGTAGACACTGCAGATGAGGATAT
- Phenotype
-
MIM: 102670
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MADCAM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59172096,rs79777104] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MADCAM1
- Gene Name
- mucosal vascular addressin cell adhesion molecule 1
There are no transcripts associated with this gene.
- Gene
- TPGS1
- Gene Name
- tubulin polyglutamylase complex subunit 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033513.2 |
|
Intron |
|
|
NP_277048.2 |
View Full Product Details