Product Details

SNP ID
rs10075211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148459974 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGAGAAAGAACCAAGTAGAAATG[C/T]TAAAGGTCAAAAACACTGCTGCAAA
Phenotype
MIM: 602164
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR4 PubMed Links

Gene Details

Gene
HTR4
Gene Name
5-hydroxytryptamine receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000870.6 Intron NP_000861.1
NM_001040169.2 Intron NP_001035259.1
NM_001040172.2 Intron NP_001035262.2
NM_001040173.2 Intron NP_001035263.1
NM_001286410.1 Intron NP_001273339.1
NM_199453.3 Intron NP_955525.1

View Full Product Details