Product Details

SNP ID
rs6090043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64086577 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTTTATTGCTGCCAAGGCCTGA[C/T]GATGAAACTTCAAGTCAAATTAGTA
Phenotype
MIM: 602548 MIM: 605071
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LKAAEAR1 PubMed Links

Gene Details

Gene
LKAAEAR1
Gene Name
LKAAEAR motif containing 1
There are no transcripts associated with this gene.

Gene
MIR6813
Gene Name
microRNA 6813
There are no transcripts associated with this gene.

Gene
OPRL1
Gene Name
opioid related nociceptin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000913.5 339 Intron NP_000904.1
NM_001200019.1 339 UTR 5 NP_001186948.1
NM_001318853.1 339 Intron NP_001305782.1
NM_001318854.1 339 Intron NP_001305783.1
NM_001318855.1 339 Intron NP_001305784.1
NM_182647.3 339 Intron NP_872588.1
XM_011528828.2 339 UTR 5 XP_011527130.1
XM_011528830.2 339 UTR 5 XP_011527132.1
XM_017027852.1 339 UTR 5 XP_016883341.1
XM_017027853.1 339 UTR 5 XP_016883342.1
XM_017027854.1 339 Intron XP_016883343.1
XM_017027855.1 339 Intron XP_016883344.1
Gene
RGS19
Gene Name
regulator of G-protein signaling 19
There are no transcripts associated with this gene.

View Full Product Details