Product Details

SNP ID
rs11905252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64081841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCTGACCCTGGTGCAGTCACT[C/G]AGGTCACTGTCTTGCTGGCCTTCTC
Phenotype
MIM: 602548 MIM: 605071 MIM: 604784
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LKAAEAR1 PubMed Links

Gene Details

Gene
LKAAEAR1
Gene Name
LKAAEAR motif containing 1
There are no transcripts associated with this gene.

Gene
MIR6813
Gene Name
microRNA 6813
There are no transcripts associated with this gene.

Gene
OPRL1
Gene Name
opioid related nociceptin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000913.5 Intron NP_000904.1
NM_001200019.1 Intron NP_001186948.1
NM_001318853.1 Intron NP_001305782.1
NM_001318854.1 Intron NP_001305783.1
NM_001318855.1 Intron NP_001305784.1
NM_182647.3 Intron NP_872588.1
XM_011528828.2 Intron XP_011527130.1
XM_011528830.2 Intron XP_011527132.1
XM_017027852.1 Intron XP_016883341.1
XM_017027853.1 Intron XP_016883342.1
XM_017027854.1 Intron XP_016883343.1
XM_017027855.1 Intron XP_016883344.1
Gene
RGS19
Gene Name
regulator of G-protein signaling 19
There are no transcripts associated with this gene.

Gene
TCEA2
Gene Name
transcription elongation factor A2
There are no transcripts associated with this gene.

View Full Product Details