Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000913.5 | Intron | NP_000904.1 | |||
NM_001200019.1 | Intron | NP_001186948.1 | |||
NM_001318853.1 | Intron | NP_001305782.1 | |||
NM_001318854.1 | Intron | NP_001305783.1 | |||
NM_001318855.1 | Intron | NP_001305784.1 | |||
NM_182647.3 | Intron | NP_872588.1 | |||
XM_011528828.2 | Intron | XP_011527130.1 | |||
XM_011528830.2 | Intron | XP_011527132.1 | |||
XM_017027852.1 | Intron | XP_016883341.1 | |||
XM_017027853.1 | Intron | XP_016883342.1 | |||
XM_017027854.1 | Intron | XP_016883343.1 | |||
XM_017027855.1 | Intron | XP_016883344.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039467.1 | Intron | NP_001034556.1 | |||
NM_005873.2 | Intron | NP_005864.1 | |||
XM_005260182.3 | Intron | XP_005260239.1 | |||
XM_005260183.2 | Intron | XP_005260240.1 | |||
XM_011528484.1 | Intron | XP_011526786.1 | |||
XM_011528485.2 | Intron | XP_011526787.1 | |||
XM_011528486.2 | Intron | XP_011526788.1 |