Product Details

SNP ID
rs2357285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:17590563 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATACATCCCATACCACTGCTGTTC[C/T]CTGGATATTTACAGAGCTCAAGACT
Phenotype
MIM: 610467
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HACD1 PubMed Links

Gene Details

Gene
HACD1
Gene Name
3-hydroxyacyl-CoA dehydratase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014241.3 Intron NP_055056.3
XM_005252641.4 Intron XP_005252698.1

View Full Product Details