Product Details

SNP ID
hCV26748561
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:14024522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCTCAAGAGAGCAAGATGTTCTC[C/G]TGTTGCTTCTCCACTTTGAGAGGTT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CDRT15P1 PubMed Links

Gene Details

Gene
CDRT15P1
Gene Name
CMT1A duplicated region transcript 15 pseudogene 1
There are no transcripts associated with this gene.

Gene
COX10-AS1
Gene Name
COX10 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details