Product Details

SNP ID
rs6632874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:16722921 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGCCTCTACTCTTTCTGAAAAAA[A/C]TAAATAAATAAAAAGAAAATCTGTA
Phenotype
MIM: 300380
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CTPS2 PubMed Links

Gene Details

Gene
CTPS2
Gene Name
CTP synthase 2
There are no transcripts associated with this gene.

Gene
SYAP1
Gene Name
synapse associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032796.3 Intron NP_116185.2

View Full Product Details