Product Details

SNP ID
rs4986049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21382614 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACCTCTGCAACTATAGAGACTGG[C/A]ATGCTTTCTGCTTGCCTGAGCACTG
Phenotype
MIM: 602323
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
KCNJ12 PubMed Links

Gene Details

Gene
KCNJ12
Gene Name
potassium voltage-gated channel subfamily J member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021012.4 Intron NP_066292.2
XM_005256625.4 Intron XP_005256682.1
XM_011523831.2 Intron XP_011522133.1

View Full Product Details