Product Details

SNP ID
rs2416298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:112976918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGAGCCGGGATGCACTGTTCCT[C/G]CTGTGGGTCCTCATCATGGAGACCA
Phenotype
MIM: 609844
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCP2 PubMed Links

Gene Details

Gene
DCP2
Gene Name
decapping mRNA 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242377.1 209 UTR 5 NP_001229306.1
NM_152624.5 209 UTR 5 NP_689837.2
XM_017009163.1 209 UTR 5 XP_016864652.1

View Full Product Details