Product Details

SNP ID
rs2592586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:177551678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTGGCAAGTGATCACAGCGTC[C/T]AAGAAGTCATAGAGATAGGGTGTGA
Phenotype
MIM: 603051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGPS PubMed Links

Gene Details

Gene
AGPS
Gene Name
alkylglycerone phosphate synthase
There are no transcripts associated with this gene.

Gene
TTC30B
Gene Name
tetratricopeptide repeat domain 30B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152517.2 1119 Silent Mutation TTA,TTG L362L NP_689730.2

View Full Product Details