Product Details

SNP ID
rs3825184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48904557 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAAACCTCATTTTCATCCTGAC[C/T]TCCCCTCCTCCGGAAATCCTGAGAC
Phenotype
MIM: 611088
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC65 PubMed Links

Gene Details

Gene
CCDC65
Gene Name
coiled-coil domain containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286957.1 Intron NP_001273886.1
NM_033124.4 Intron NP_149115.2

View Full Product Details