Product Details

SNP ID
rs35034312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48966364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGCTCTCCTGAGAGGCGACGGG[A/G]ACGCAGACGGGGCTGGAAGGCTCCA
Phenotype
MIM: 103190 MIM: 601906
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARF3 PubMed Links

Gene Details

Gene
ARF3
Gene Name
ADP ribosylation factor 3
There are no transcripts associated with this gene.

Gene
WNT10B
Gene Name
Wnt family member 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003394.3 743 Missense Mutation CCC,TCC P301S NP_003385.2
XM_011538722.1 743 Missense Mutation CCC,TCC P179S XP_011537024.1
XM_011538724.1 743 UTR 3 XP_011537026.1
XM_017019919.1 743 Missense Mutation CCC,TCC P179S XP_016875408.1

View Full Product Details